Product Details

SNP ID
rs186619470
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:248626440 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAGTGGTGAAGGCCTTTTTGCGA[A/C]CTTCAGCAGAGGGCATGCGGTGGAT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR2T10 PubMed Links

Gene Details

Gene
OR2T10
Gene Name
olfactory receptor family 2 subfamily T member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004693.1 689 Intron NP_001004693.1
XM_017000273.1 689 Intron XP_016855762.1
Gene
OR2T11
Gene Name
olfactory receptor family 2 subfamily T member 11 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001964.1 689 Missense Mutation GGT,GTT G230V NP_001001964.1

View Full Product Details