Product Details

SNP ID
rs186308074
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:57248556 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCAGATGACGAGTTCCAGTTATT[A/T]CAGAGAAATTTCATGGACAAGTACT
Phenotype
MIM: 615407 MIM: 600340 MIM: 616585
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ARL2BP PubMed Links

Gene Details

Gene
ARL2BP
Gene Name
ADP ribosylation factor like GTPase 2 binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012106.3 362 Missense Mutation TTA,TTT L40F NP_036238.1
XM_011522977.1 362 Intron XP_011521279.1
Gene
PLLP
Gene Name
plasmolipin
There are no transcripts associated with this gene.

Gene
RSPRY1
Gene Name
ring finger and SPRY domain containing 1
There are no transcripts associated with this gene.

View Full Product Details