Product Details

SNP ID
rs186417780
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:109714421 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATAATGACACCTGCATGCGGCTC[A/G]TTGGCTGTCCAGATATGGTTTACAA
Phenotype
MIM: 611652
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PLA2G12A PubMed Links

Gene Details

Gene
PLA2G12A
Gene Name
phospholipase A2 group XIIA
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030821.4 803 Nonsense Mutation CGA,TGA R176* NP_110448.2

View Full Product Details