Product Details

SNP ID
rs186835351
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12624669 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTAGGGGAAAAATGGGAAGACCC[A/G]AATGTTGAAGATCAACACAAAAACC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF791 PubMed Links

Gene Details

Gene
ZNF791
Gene Name
zinc finger protein 791
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153358.2 282 Silent Mutation CCA,CCG P50P NP_699189.2
XM_017026412.1 282 Silent Mutation CCA,CCG P18P XP_016881901.1

View Full Product Details