Product Details

SNP ID
rs186832952
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51480410 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAACCCTGGGGATCAAGGGATAT[A/C]GGACTGTCGTGGCCCTGGATAAGGT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CEACAM18 PubMed Links

Gene Details

Gene
CEACAM18
Gene Name
carcinoembryonic antigen related cell adhesion molecule 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278392.1 130 Silent Mutation AGG,CGG R44R NP_001265321.1
XM_011527258.2 130 Silent Mutation AGG,CGG R44R XP_011525560.1

View Full Product Details