Product Details

SNP ID
rs187222480
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:121641622 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCACAGAGGGGACCACCCCCTGAC[A/G]CCCGGCAGCCACTATGCCTAGGCCC
Phenotype
MIM: 610277
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MORN3 PubMed Links

Gene Details

Gene
MORN3
Gene Name
MORN repeat containing 3
There are no transcripts associated with this gene.

Gene
ORAI1
Gene Name
ORAI calcium release-activated calcium modulator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032790.3 1078 Silent Mutation ACA,ACG T295T NP_116179.2

View Full Product Details