Product Details

SNP ID
rs187232235
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:110057270 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGCAGGGAGGATGTGCCTTTCA[A/G]CAAAGTTCCTCTCCCCCTTACAGCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C12orf76 PubMed Links

Gene Details

Gene
C12orf76
Gene Name
chromosome 12 open reading frame 76
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207435.2 251 Missense Mutation GCT,GTT A73V NP_997318.1
XM_017019297.1 251 Missense Mutation GCT,GTT A84V XP_016874786.1

View Full Product Details