Product Details

SNP ID
rs186877232
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:80458673 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGTGGAAGAGAAAATAGTTGATT[A/G]GAAAAGCCCACAGTCTTTTAGATTC
Phenotype
MIM: 139395
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GNAT3 PubMed Links

Gene Details

Gene
GNAT3
Gene Name
G protein subunit alpha transducin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011516124.2 Intron XP_011514426.1

View Full Product Details