Product Details

SNP ID
rs186878325
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:30198456 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCTTTTTCCTCTCGGTGAGTGTG[C/T]TACACAGCATTCAAGCGGATGTCCC
Phenotype
MIM: 182138
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC6A4 PubMed Links

Gene Details

Gene
SLC6A4
Gene Name
solute carrier family 6 member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001045.5 2469 Silent Mutation TAA,TAG *631* NP_001036.1

View Full Product Details