Product Details

SNP ID
rs187659169
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:50375904 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGGCAATATTAAGAAAACCTTAT[C/G]ATTAAGAAACTGTGGTAAGTAAAAC
Phenotype
MIM: 616673
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SKA1 PubMed Links

Gene Details

Gene
SKA1
Gene Name
spindle and kinetochore associated complex subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039535.2 258 Nonsense Mutation TCA,TGA S25* NP_001034624.1
NM_145060.3 258 Nonsense Mutation TCA,TGA S25* NP_659497.1

View Full Product Details