Product Details

SNP ID
rs187435008
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:13922080 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAGAAGGAGTTGAACACCTCCCTC[A/G]CCGTGTAACAAATGAAATCACAAGC
Phenotype
MIM: 133520
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ERCC4 PubMed Links

Gene Details

Gene
ERCC4
Gene Name
ERCC excision repair 4, endonuclease catalytic subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005236.2 232 Missense Mutation CAC,CGC H86R NP_005227.1
XM_011522424.2 232 Missense Mutation CAC,CGC H86R XP_011520726.1
XM_011522427.1 232 Intron XP_011520729.1
XM_017023043.1 232 UTR 5 XP_016878532.1

View Full Product Details