Product Details

SNP ID
rs187446768
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:9213906 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTGAGGCCCTGATTTGTCATGGA[C/T]AAGAGTCGGCCCTGCTGAGGAGTCT
Phenotype
MIM: 611538
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR7D4 PubMed Links

Gene Details

Gene
OR7D4
Gene Name
olfactory receptor family 7 subfamily D member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005191.2 Intron NP_001005191.1

View Full Product Details