Product Details

SNP ID
rs187539558
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:34710020 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTACCTTGGGTCCTGGGGATGCCA[A/G]AGGCCAGAACCAGGATAAGGAGGCT
Phenotype
MIM: 602737
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCL21 PubMed Links

Gene Details

Gene
CCL21
Gene Name
C-C motif chemokine ligand 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002989.3 148 Missense Mutation TCT,TTT S16F NP_002980.1
XM_011518004.2 148 Missense Mutation TCT,TTT S16F XP_011516306.1

View Full Product Details