Product Details

SNP ID
rs187555579
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:14446970 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAGAGGGGCTCTGCACAGGAATC[A/G]GTCTGCTCAAGCTCAGTGTCGGAAA
Phenotype
MIM: 604212
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
PARN PubMed Links

Gene Details

Gene
PARN
Gene Name
poly(A)-specific ribonuclease
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134477.2 1845 Silent Mutation ACC,ACT T533T NP_001127949.1
NM_001242992.1 1845 Silent Mutation ACC,ACT T548T NP_001229921.1
NM_002582.3 1845 Silent Mutation ACC,ACT T594T NP_002573.1
XM_011522510.2 1845 Intron XP_011520812.1
XM_011522511.2 1845 Intron XP_011520813.1
XM_011522513.2 1845 Intron XP_011520815.1
XM_011522514.1 1845 Intron XP_011520816.1
XM_017023258.1 1845 Intron XP_016878747.1
XM_017023259.1 1845 Intron XP_016878748.1
XM_017023260.1 1845 Intron XP_016878749.1

View Full Product Details