Product Details

SNP ID
rs187883081
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:79171436 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTACATCAGCATTGCTGGCTGGCT[G/T]ATCATCTGCCTTGCCTGTGTACTCT
Phenotype
MIM: 300903
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GPR174 PubMed Links

Gene Details

Gene
GPR174
Gene Name
G protein-coupled receptor 174
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032553.1 465 Silent Mutation CTG,CTT L143L NP_115942.1

View Full Product Details