Product Details

SNP ID
rs188191103
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:124869529 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCAGCAGTACTGGAATGCGTGCC[C/T]CCCCGCGGCCACCCGGAGCCTTCCG
Phenotype
MIM: 608630
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ROBO3 PubMed Links

Gene Details

Gene
ROBO3
Gene Name
roundabout guidance receptor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022370.3 736 Silent Mutation CCC,CCT P189P NP_071765.2
XM_017018122.1 736 Silent Mutation CCC,CCT P167P XP_016873611.1

View Full Product Details