Product Details

SNP ID
rs188345683
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:45999655 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGGGTGAGAGTCAGGCGACCTCA[C/T]GCCAAGTGTAGAAAGGGGCAGATGG
Phenotype
MIM: 157140 MIM: 607067
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAPT PubMed Links

Gene Details

Gene
MAPT
Gene Name
microtubule associated protein tau
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001123066.3 406 Intron NP_001116538.2
NM_001123067.3 406 Intron NP_001116539.1
NM_001203251.1 406 Intron NP_001190180.1
NM_001203252.1 406 Intron NP_001190181.1
NM_005910.5 406 Intron NP_005901.2
NM_016834.4 406 Intron NP_058518.1
NM_016835.4 406 Intron NP_058519.3
NM_016841.4 406 Intron NP_058525.1
XM_005257362.4 406 Intron XP_005257419.1
XM_005257364.4 406 Intron XP_005257421.1
XM_005257365.4 406 Intron XP_005257422.1
XM_005257366.3 406 Intron XP_005257423.1
XM_005257367.4 406 Intron XP_005257424.1
XM_005257368.4 406 Intron XP_005257425.1
XM_005257369.4 406 Intron XP_005257426.1
XM_005257370.4 406 Intron XP_005257427.1
XM_005257371.4 406 Intron XP_005257428.1
Gene
STH
Gene Name
saitohin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007532.2 406 Missense Mutation CGC,TGC R126C NP_001007533.1

View Full Product Details