Product Details
- SNP ID
-
rs188238762
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:1528907 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTGTGCCTCCCTCAGGCCCCTTCA[C/T]GCCTAAAGAGGCCTCGGGTCCAGGA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PLK5
PubMed Links
Gene Details
- Gene
- PLK5
- Gene Name
- polo like kinase 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001243079.1 |
1054 |
Missense Mutation |
ACG,ATG |
T113M |
NP_001230008.1 |
View Full Product Details