Product Details

SNP ID
rs188043956
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:106761508 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAGTTGAGTTTCTCTGTAAATCT[A/G]TTTACCCTAAAGTAGAGAGAGGGTG
Phenotype
MIM: 609747 MIM: 605609
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABRA PubMed Links

Gene Details

Gene
ABRA
Gene Name
actin binding Rho activating protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139166.4 729 Silent Mutation AAC,AAT N225N NP_631905.1
Gene
OXR1
Gene Name
oxidation resistance 1
There are no transcripts associated with this gene.

View Full Product Details