Product Details

SNP ID
rs188433430
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:53290028 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCAAATCCCATTCGTATAGCTTCT[C/T]GTAGCATAGGATTAGGGAAGATGGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BIRC8 PubMed Links

Gene Details

Gene
BIRC8
Gene Name
baculoviral IAP repeat containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033341.4 Intron NP_203127.3
Gene
FAM90A27P
Gene Name
family with sequence similarity 90, member A27, pseudogene
There are no transcripts associated with this gene.

View Full Product Details