Product Details

SNP ID
rs188691406
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:29007916 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATAGATATGATGGCTGTAGATCCA[C/G]AGTCTCGTGAAGATGAGGTCAACTG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RWDD2B PubMed Links

Gene Details

Gene
RWDD2B
Gene Name
RWD domain containing 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320724.1 762 Silent Mutation CTC,CTG L161L NP_001307653.1
NM_016940.2 762 Silent Mutation CTC,CTG L190L NP_058636.1

View Full Product Details