Product Details

SNP ID
rs188827537
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:69121125 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGTAGCGAGCCTCGATCTCCCC[C/T]TGTAGCTCCATCAGCACCCATTCTG
Phenotype
MIM: 613202 MIM: 602428
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHTF8 PubMed Links

Gene Details

Gene
CHTF8
Gene Name
chromosome transmission fidelity factor 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039690.3 Intron NP_001034779.1
NM_001040146.3 Intron NP_001035236.1
XM_011523167.1 Intron XP_011521469.1
XM_011523168.1 Intron XP_011521470.1
XM_017023370.1 Intron XP_016878859.1
Gene
HAS3
Gene Name
hyaluronan synthase 3
There are no transcripts associated with this gene.

View Full Product Details