Product Details

SNP ID
rs188816777
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:50288241 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCACACGGCTTCGAGCCTTGGTCC[A/G]GGCTTTGAAGGCAGCAGCATTGTAC
Phenotype
MIM: 604038 MIM: 602725 MIM: 607073
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HYAL3 PubMed Links

Gene Details

Gene
HYAL3
Gene Name
hyaluronoglucosaminidase 3
There are no transcripts associated with this gene.

Gene
IFRD2
Gene Name
interferon related developmental regulator 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006764.4 1600 Missense Mutation CGG,TGG R491W NP_006755.4
Gene
LSMEM2
Gene Name
leucine rich single-pass membrane protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304385.1 1600 Intron NP_001291314.1
NM_153215.2 1600 Intron NP_694947.1
XM_006712979.3 1600 Intron XP_006713042.1
XM_006712980.3 1600 Intron XP_006713043.1
XM_011533370.2 1600 Intron XP_011531672.1
Gene
NAT6
Gene Name
N-acetyltransferase 6
There are no transcripts associated with this gene.

View Full Product Details