Product Details

SNP ID
rs188767367
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:49905823 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCACAGCGGGAAGTGGGCGTGGT[A/G]GAGGGCTTGGATTTGGAGTGAAGGT
Phenotype
MIM: 607662
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC105372653 PubMed Links

Gene Details

Gene
LOC105372653
Gene Name
uncharacterized LOC105372653
There are no transcripts associated with this gene.

Gene
SPATA2
Gene Name
spermatogenesis associated 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135773.1 1582 Silent Mutation TCC,TCT S453S NP_001129245.1
NM_006038.3 1582 Silent Mutation TCC,TCT S453S NP_006029.1
XM_006723894.1 1582 Silent Mutation TCC,TCT S453S XP_006723957.1
XM_011529116.1 1582 Silent Mutation TCC,TCT S453S XP_011527418.1

View Full Product Details