Product Details
- SNP ID
-
rs188560839
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:92298208 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCCTTCCAGGACTGTAATTCTAGA[C/T]AGGTAACATGTCTATTTTAGGAAAT
- Phenotype
-
MIM: 600709
MIM: 611534
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
IARS
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3831004] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- IARS
- Gene Name
- isoleucyl-tRNA synthetase
There are no transcripts associated with this gene.
- Gene
- MIR3651
- Gene Name
- microRNA 3651
There are no transcripts associated with this gene.
- Gene
- NOL8
- Gene Name
- nucleolar protein 8
- Gene
- SNORA84
- Gene Name
- small nucleolar RNA, H/ACA box 84
There are no transcripts associated with this gene.
View Full Product Details