Product Details

SNP ID
rs188614646
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:248239015 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCCACCCACACCTTCCTTTTTTC[C/T]CTGGTCCTGGGCATCTTCTCACTGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR2M4 PubMed Links

Gene Details

Gene
OR2M4
Gene Name
olfactory receptor family 2 subfamily M member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017504.1 87 Silent Mutation TCC,TCT S29S NP_059974.1

View Full Product Details