Product Details

SNP ID
rs188625148
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:169778861 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATAAAGATGGAAGATTTTGCTTT[C/T]ATTGCTAATCCTTCTTCTACAGAGA
Phenotype
MIM: 608534 MIM: 606042
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACTRT3 PubMed Links

Gene Details

Gene
ACTRT3
Gene Name
actin related protein T3
There are no transcripts associated with this gene.

Gene
MYNN
Gene Name
myoneurin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185118.1 1547 Silent Mutation TTC,TTT F120F NP_001172047.1
NM_001185119.1 1547 Silent Mutation TTC,TTT F120F NP_001172048.1
NM_018657.4 1547 Silent Mutation TTC,TTT F120F NP_061127.1
XM_005247621.4 1547 Silent Mutation TTC,TTT F120F XP_005247678.1
XM_005247622.3 1547 Intron XP_005247679.1
XM_005247624.3 1547 Silent Mutation TTC,TTT F6F XP_005247681.1
XM_011512987.1 1547 Missense Mutation TTC,TTT F120F XP_011511289.1
XM_017006864.1 1547 Silent Mutation TTC,TTT F120F XP_016862353.1
XM_017006865.1 1547 Silent Mutation TTC,TTT F6F XP_016862354.1
XM_017006866.1 1547 Silent Mutation TTC,TTT F6F XP_016862355.1
XM_017006867.1 1547 Intron XP_016862356.1
XM_017006868.1 1547 Intron XP_016862357.1

View Full Product Details