Product Details

SNP ID
rs188400743
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:73213273 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATTCCAGTGCATCACCTGAGAAA[A/T]ATAATACTGATCTTGGAATTATGTA
Phenotype
MIM: 300026
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
NAP1L2 PubMed Links

Gene Details

Gene
NAP1L2
Gene Name
nucleosome assembly protein 1 like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021963.3 1602 Missense Mutation TAT,TTT Y407F NP_068798.1

View Full Product Details