Product Details

SNP ID
rs189411535
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:27980179 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGATGTTTGTTGGGAAATCACAAT[A/T]AACCAGATGATGAGGAGTGAATTTC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DCAF8L1 PubMed Links

Gene Details

Gene
DCAF8L1
Gene Name
DDB1 and CUL4 associated factor 8 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017930.1 1271 Missense Mutation AAT,TAT N386Y NP_001017930.1

View Full Product Details