Product Details

SNP ID
rs188911161
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24207261 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGTGGGTTGTAGCCCTTCTTAC[C/T]GCCAAGTGAACTGCTTTTCCCACCT
Phenotype
MIM: 610051 MIM: 603171 MIM: 610711
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHMP4A PubMed Links

Gene Details

Gene
CHMP4A
Gene Name
charged multivesicular body protein 4A
There are no transcripts associated with this gene.

Gene
MDP1
Gene Name
magnesium dependent phosphatase 1
There are no transcripts associated with this gene.

Gene
NEDD8
Gene Name
neural precursor cell expressed, developmentally down-regulated 8
There are no transcripts associated with this gene.

Gene
NEDD8-MDP1
Gene Name
NEDD8-MDP1 readthrough
There are no transcripts associated with this gene.

Gene
TSSK4
Gene Name
testis specific serine kinase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184739.1 790 Missense Mutation CGC,TGC R196C NP_001171668.1
NM_001308067.1 790 Missense Mutation CGC,TGC R110C NP_001294996.1
NM_174944.3 790 Missense Mutation CGC,TGC R186C NP_777604.2
XM_011536663.2 790 Missense Mutation CGC,TGC R120C XP_011534965.1
XM_017021226.1 790 Missense Mutation CGC,TGC R186C XP_016876715.1

View Full Product Details