Product Details

SNP ID
rs188943586
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:25442718 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAAAGGTAGGAATAGAATGGGGG[A/G]AAACCTTATAGATAATATTCACTTT
Phenotype
MIM: 610301
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM57 PubMed Links

Gene Details

Gene
TMEM57
Gene Name
transmembrane protein 57
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282564.1 Intron NP_001269493.1
NM_018202.5 Intron NP_060672.2
XM_005245931.2 Intron XP_005245988.1
XM_011541704.2 Intron XP_011540006.1

View Full Product Details