Product Details

SNP ID
rs189560074
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:11668685 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGATGCTCCCCACTCCAAAGCAGC[C/T]CTGGACAGCATTAACGAGCTGCCCG
Phenotype
MIM: 609111 MIM: 605647 MIM: 604094
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FBXO44 PubMed Links

Gene Details

Gene
FBXO44
Gene Name
F-box protein 44
There are no transcripts associated with this gene.

Gene
FBXO6
Gene Name
F-box protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018438.5 133 Silent Mutation GCC,GCT A9A NP_060908.1
XM_005263448.4 133 Silent Mutation GCC,GCT A9A XP_005263505.1
XM_005263449.4 133 Silent Mutation GCC,GCT A9A XP_005263506.1
XM_005263451.4 133 Silent Mutation GCC,GCT A9A XP_005263508.1
XM_006710571.3 133 Silent Mutation GCC,GCT A9A XP_006710634.1
XM_011541232.2 133 Silent Mutation GCC,GCT A9A XP_011539534.1
XM_017001007.1 133 Silent Mutation GCC,GCT A9A XP_016856496.1
Gene
MAD2L2
Gene Name
MAD2 mitotic arrest deficient-like 2 (yeast)
There are no transcripts associated with this gene.

View Full Product Details