Product Details

SNP ID
rs189666922
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:34794460 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCCGCTACTGGGTGCTGCAGTTC[C/T]TCTTCGTCAGCACACCCACCCTGGT
Phenotype
MIM: 121012 MIM: 603324
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GJA4 PubMed Links

Gene Details

Gene
GJA4
Gene Name
gap junction protein alpha 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002060.2 504 Missense Mutation CTC,TTC L83F NP_002051.2
XM_005270750.2 504 Missense Mutation CTC,TTC L83F XP_005270807.1
XM_017001043.1 504 Missense Mutation CTC,TTC L83F XP_016856532.1
Gene
GJB3
Gene Name
gap junction protein beta 3
There are no transcripts associated with this gene.

View Full Product Details