Product Details

SNP ID
rs190519121
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:151095212 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTTTTCTACAACGTACATTTCTC[C/T]GTTTGCTGCAAGTCAGGCACTGTCT
Phenotype
MIM: 616813
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AGAP3 PubMed Links

Gene Details

Gene
AGAP3
Gene Name
ArfGAP with GTPase domain, ankyrin repeat and PH domain 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042535.3 Intron NP_001036000.1
NM_001281300.1 Intron NP_001268229.1
NM_001308304.1 Intron NP_001295233.1
NM_001308305.1 Intron NP_001295234.1
NM_031946.6 Intron NP_114152.3
XM_005249943.2 Intron XP_005250000.1
XM_011515780.1 Intron XP_011514082.1
XM_017011732.1 Intron XP_016867221.1
XM_017011733.1 Intron XP_016867222.1
XM_017011734.1 Intron XP_016867223.1
XM_017011735.1 Intron XP_016867224.1

View Full Product Details