Product Details

SNP ID
rs190506495
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:156298781 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGCATTGCTCTTTCAGGGTATAC[A/C]CTGCAGTGTGATGTTGGCAAAAACA
Phenotype
MIM: 610962 MIM: 615531
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GLMP PubMed Links

Gene Details

Gene
GLMP
Gene Name
glycosylated lysosomal membrane protein
There are no transcripts associated with this gene.

Gene
SMG5
Gene Name
SMG5, nonsense mediated mRNA decay factor
There are no transcripts associated with this gene.

Gene
TMEM79
Gene Name
transmembrane protein 79
There are no transcripts associated with this gene.

Gene
VHLL
Gene Name
von Hippel-Lindau tumor suppressor like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004319.2 857 Missense Mutation GGT,TGT G137C NP_001004319.1

View Full Product Details