Product Details

SNP ID
rs190418341
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:144074841 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGGGGCAGTGTCTGTGCTGGTG[A/G]GAGCCTTCTTTTCCACTGTAATATC
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
OR2A25 PubMed Links
Additional Information
For this assay, SNP(s) [rs2961135] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
OR2A25
Gene Name
olfactory receptor family 2 subfamily A member 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004488.1 622 Missense Mutation AGA,GGA R208G NP_001004488.1

View Full Product Details