Product Details

SNP ID
rs190430069
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:152990553 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTGCCTGTCTACCGGAGGCCCCC[C/T]GGCCTCTCCCTGATGGCTTCTCCTT
Phenotype
MIM: 300916
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PNMA5 PubMed Links

Gene Details

Gene
PNMA5
Gene Name
paraneoplastic Ma antigen family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001103150.1 1385 Missense Mutation CAG,CGG Q349R NP_001096620.1
NM_001103151.1 1385 Missense Mutation CAG,CGG Q349R NP_001096621.1
NM_001184924.1 1385 Missense Mutation CAG,CGG Q349R NP_001171853.1
NM_052926.2 1385 Missense Mutation CAG,CGG Q349R NP_443158.1
XM_017029252.1 1385 Missense Mutation CAG,CGG Q349R XP_016884741.1
XM_017029253.1 1385 Missense Mutation CAG,CGG Q349R XP_016884742.1

View Full Product Details