Product Details

SNP ID
rs190147459
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:32209183 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGCTCCCAGCTGGTGGTGACTCA[A/G]CGCCGCTTCCCCACCATGGAGGCCT
Phenotype
MIM: 610162
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC28B PubMed Links

Gene Details

Gene
CCDC28B
Gene Name
coiled-coil domain containing 28B
There are no transcripts associated with this gene.

Gene
DCDC2B
Gene Name
doublecortin domain containing 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099434.1 371 Silent Mutation CAA,CAG Q30Q NP_001092904.1
XM_017000413.1 371 Silent Mutation CAA,CAG Q30Q XP_016855902.1
XM_017000414.1 371 Intron XP_016855903.1
Gene
IQCC
Gene Name
IQ motif containing C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160042.1 371 Intron NP_001153514.1
NM_018134.2 371 Intron NP_060604.2
Gene
TMEM234
Gene Name
transmembrane protein 234
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019118.4 371 Intron NP_061991.3
XM_011541795.2 371 Intron XP_011540097.1
XM_017001812.1 371 Intron XP_016857301.1
XM_017001813.1 371 Intron XP_016857302.1
XM_017001814.1 371 Intron XP_016857303.1
XM_017001815.1 371 Intron XP_016857304.1
XM_017001816.1 371 Intron XP_016857305.1
XM_017001817.1 371 Intron XP_016857306.1
XM_017001818.1 371 Intron XP_016857307.1
XM_017001819.1 371 Intron XP_016857308.1
XM_017001820.1 371 Intron XP_016857309.1
XM_017001821.1 371 Intron XP_016857310.1
XM_017001822.1 371 Intron XP_016857311.1
XM_017001823.1 371 Intron XP_016857312.1
XM_017001824.1 371 Intron XP_016857313.1
XM_017001825.1 371 Intron XP_016857314.1
XM_017001826.1 371 Intron XP_016857315.1
XM_017001827.1 371 Intron XP_016857316.1
XM_017001828.1 371 Intron XP_016857317.1
XM_017001829.1 371 Intron XP_016857318.1
XM_017001830.1 371 Intron XP_016857319.1
XM_017001831.1 371 Intron XP_016857320.1

View Full Product Details