Product Details

SNP ID
rs190126440
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:50193286 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGGATTATCCACCAGGACGGGTA[C/G]TCGCTGGAAGAGTGCCTCGAGTTTA
Phenotype
MIM: 139330 MIM: 601124
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GNAT1 PubMed Links

Gene Details

Gene
GNAT1
Gene Name
G protein subunit alpha transducin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000172.3 287 Nonsense Mutation TAC,TAG Y57* NP_000163.2
NM_144499.2 287 Nonsense Mutation TAC,TAG Y57* NP_653082.1
Gene
SEMA3F
Gene Name
semaphorin 3F
There are no transcripts associated with this gene.

View Full Product Details