Product Details

SNP ID
rs190215470
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:115202164 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCATTCCATCAGAGGCATTCATTT[A/G]CCTTAAGAGGTTTAGAATATTACAG
Phenotype
MIM: 602031
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PGGT1B PubMed Links

Gene Details

Gene
PGGT1B
Gene Name
protein geranylgeranyltransferase type I subunit beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005023.3 11296 Intron NP_005014.2
XM_005272020.3 11296 UTR 3 XP_005272077.2
XM_011543490.2 11296 Intron XP_011541792.1
XM_011543492.2 11296 Intron XP_011541794.1

View Full Product Details