Product Details

SNP ID
rs190313223
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:67992921 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATACCTGTCGTCTAAATCCCATTC[A/G]TGTCATTGTTTCCCATCGCTCCCAA
Phenotype
MIM: 601819
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BPTF PubMed Links

Gene Details

Gene
BPTF
Gene Name
bromodomain PHD finger transcription factor
There are no transcripts associated with this gene.

Gene
C17orf58
Gene Name
chromosome 17 open reading frame 58
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181655.3 Intron NP_858041.2
NM_181656.4 Intron NP_858042.2

View Full Product Details