Product Details

SNP ID
rs190632628
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:28349181 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACGATCAGATGAGATTTCATCGC[A/G]GGTGCTGGCTGCTGGGGCTCGCTGA
Phenotype
MIM: 601459 MIM: 609494
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PNOC PubMed Links

Gene Details

Gene
PNOC
Gene Name
prepronociceptin
There are no transcripts associated with this gene.

Gene
ZNF395
Gene Name
zinc finger protein 395
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018660.2 1500 Silent Mutation CCC,CCT P458P NP_061130.1

View Full Product Details