Product Details

SNP ID
rs190908366
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:157463152 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTCCTGTGCCAGATTGTCAATGA[C/T]CTCAGCCCTGAGGTGCCCAGCAATG
Phenotype
MIM: 609383
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC102724404 PubMed Links

Gene Details

Gene
LOC102724404
Gene Name
uncharacterized LOC102724404
There are no transcripts associated with this gene.

Gene
NIPAL4
Gene Name
NIPA like domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099287.1 398 Silent Mutation GAC,GAT D94D NP_001092757.1
NM_001172292.1 398 Silent Mutation GAC,GAT D94D NP_001165763.1
XM_011534552.1 398 UTR 5 XP_011532854.1

View Full Product Details