Product Details

SNP ID
rs190903361
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:81992288 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATTAGGATCTGTGGAGTGTGTTT[G/T]CATAGACCCCAGAACGAGGCACGGT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SPATA31D1 PubMed Links

Gene Details

Gene
SPATA31D1
Gene Name
SPATA31 subfamily D member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001670.2 1843 Missense Mutation TTG,TTT L606F NP_001001670.1
XM_017014710.1 1843 Missense Mutation TTG,TTT L569F XP_016870199.1

View Full Product Details