Product Details

SNP ID
rs190940468
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48904438 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCGAGAGAAGCTGCAGGCTGCCA[A/G]TGCGGAGGACATCAAGGTGCGGCTG
Phenotype
MIM: 601323 MIM: 602309
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NUCB1 PubMed Links

Gene Details

Gene
NUCB1
Gene Name
nucleobindin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006184.5 561 Missense Mutation AAT,AGT N76S NP_006175.2
XM_017026845.1 561 Missense Mutation AAT,AGT N76S XP_016882334.1
Gene
NUCB1-AS1
Gene Name
NUCB1 antisense RNA 1
There are no transcripts associated with this gene.

Gene
TULP2
Gene Name
tubby like protein 2
There are no transcripts associated with this gene.

View Full Product Details