Product Details

SNP ID
rs191021241
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:95406125 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACTGAAAATATAAAAACCTCTGA[C/G]GAGGGAGGATTGTTCGGGTCATACA
Phenotype
MIM: 602447 MIM: 602720
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PON2 PubMed Links

Gene Details

Gene
PON2
Gene Name
paraoxonase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000305.2 896 Silent Mutation TCC,TCG S300S NP_000296.2
NM_001018161.1 896 Silent Mutation TCC,TCG S288S NP_001018171.1
XM_005250453.1 896 Silent Mutation TCC,TCG S232S XP_005250510.1
XM_017012357.1 896 Silent Mutation TCC,TCG S230S XP_016867846.1
XM_017012358.1 896 Silent Mutation TCC,TCG S214S XP_016867847.1
Gene
PON3
Gene Name
paraoxonase 3
There are no transcripts associated with this gene.

View Full Product Details