Product Details
- SNP ID
-
rs190686601
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:11352739 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAATCGAATATCATAGTCTGTTAA[C/T]GTCTGGTACAATTGCTTGAAGTCTG
- Phenotype
-
MIM: 615136
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CSNK2A3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2896585] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CSNK2A3
- Gene Name
- casein kinase 2 alpha 3
- Gene
- GALNT18
- Gene Name
- polypeptide N-acetylgalactosaminyltransferase 18
View Full Product Details