Product Details

SNP ID
rs190727318
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:2933689 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACTGTGTGATCTCACATGCTTTT[G/T]AAAGTACTGAGCGTGGCTGAAGGCT
Phenotype
MIM: 194551
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF77 PubMed Links

Gene Details

Gene
ZNF77
Gene Name
zinc finger protein 77
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021217.2 1594 Missense Mutation AAA,CAA K480Q NP_067040.1
XM_017027081.1 1594 Missense Mutation AAA,CAA K300Q XP_016882570.1
XM_017027082.1 1594 Missense Mutation AAA,CAA K300Q XP_016882571.1

View Full Product Details