Product Details

SNP ID
rs190773126
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:32217011 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTCTCAATTTCCTCCTCTAGGCG[G/T]GAGAATCACTCCTGTGTTACCTTAC
Phenotype
MIM: 604866 MIM: 607491
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PLAGL2 PubMed Links

Gene Details

Gene
PLAGL2
Gene Name
PLAG1 like zinc finger 2
There are no transcripts associated with this gene.

Gene
POFUT1
Gene Name
protein O-fucosyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015352.1 595 Intron NP_056167.1
NM_172236.1 595 Silent Mutation CGG,CGT R182R NP_758436.1

View Full Product Details