Product Details

SNP ID
hCV186666747
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:44697219 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCCACCATGTCCGTCTGCTCCAG[C/T]GACCTGAGCTATGGCAGCCGCGTCT
Phenotype
MIM: 612920
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRTAP10-12 PubMed Links

Gene Details

Gene
KRTAP10-12
Gene Name
keratin associated protein 10-12
There are no transcripts associated with this gene.

Gene
TSPEAR
Gene Name
thrombospondin type laminin G domain and EAR repeats
There are no transcripts associated with this gene.

View Full Product Details